missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | Cytosolic Sulfotransferase 2A1/SULT2A1 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618847
|
Novus Biologicals
NBP2-99792-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651849
|
Novus Biologicals
NBP2-99792-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISASpecifications
| Cytosolic Sulfotransferase 2A1/SULT2A1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 6822 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1 | |
| Produced in rabbits immunized with purified, recombinant Human Cytosolic Sulfotransferase 2A1/SULT2A1 (Accession#: NP_003158.2; Ser 2-Glu 285) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title