missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, DyLight 755, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Cytosolic Sulfotransferase 2A1/SULT2A1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 2A1/SULT2A1 (NP_003158.2; Ser 2-Glu 285). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?