missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18334676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334676 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334676 Supplier Novus Biologicals Supplier No. NBP299792AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 2A1/SULT2A1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 2A1/SULT2A1 (NP_003158.2; Ser 2-Glu 285).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.