missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 1E1/SULT1E1 Rabbit anti-Human, Clone: 27, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89871-50UL
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Monoclonal specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| Cytosolic Sulfotransferase 1E1/SULT1E1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 27 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4 | |
| Recombinant Human Cytosolic Sulfotransferase 1E1/SULT1E1 Protein | |
| 50 μg | |
| Breast Cancer, Stem Cell Markers | |
| 6783 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction