missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody (SULT1E1/7124), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26733-20ug
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Cytosolic Sulfotransferase 1E1/SULT1E1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4 | |
| Recombinant fragment (around aa100-294) of human Cytosolic Sulfotransferase 1E1/SULT1E1 protein (exact sequence is proprietary) | |
| 20 μg | |
| Breast Cancer, Stem Cell Markers | |
| 6783 | |
| Store at 4°C. | |
| IgG2 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| SULT1E1/7124 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction