missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic Sulfotransferase 1C2/SULT1C2 Antibody [PE], Novus Biologicals™

Product Code. 30735194 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735194 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735194 Supplier Novus Biologicals Supplier No. AF7458P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Cytosolic Sulfotransferase 1C2/SULT1C2 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 1C2/SULT1C2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 1C2/SULT1C2
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias cytosolic, 1C, member 1, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.2, humSULTC2, ST1C2, Sulfotransferase 1C1, sulfotransferase family, cytosolic, 1C, member 2, SULT1C#1
Host Species Sheep
Immunogen E. coli-derived recombinant human Cytosolic Sulfotransferase 1C2/SULT1C2, Ala2-Leu296, predicted, Accession # O00338
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6819
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.