Learn More
Cytosolic Sulfotransferase 1B1/SULT1B1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99836PCP
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1B1/SULT1B1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 1B1/SULT1B1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| PBS | |
| RUO | |
| Primary | |
| Store at 4C in the dark. |