missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 1B1/SULT1B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99836-50ul
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1B1/SULT1B1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 1B1/SULT1B1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Cytosolic Sulfotransferase 1B1/SULT1B1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B2, Thyroid hormone sulfotransferase | |
| Produced in rabbits immunized with purified, recombinant Human Cytosolic Sulfotransferase 1B1/SULT1B1 (Accession#: NP_055280.2; Leu 2-Ile 296) | |
| 50 μg | |
| metabolism | |
| 27284 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction