missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic Sulfotransferase 1A1/SULT1A1 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18393886 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393886 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393886 Supplier Novus Biologicals Supplier No. NBP299855IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cytosolic Sulfotransferase 1A1/SULT1A1 Polyclonal specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 1A1/SULT1A1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 1A1/SULT1A1 (P50225-1; Glu 2-Leu 295).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6817
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.