missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic Sulfotransferase 1A1/SULT1A1 Antibody (034), DyLight 650, Novus Biologicals™

Product Code. 18376250 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376250 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376250 Supplier Novus Biologicals Supplier No. NBP289950C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytosolic Sulfotransferase 1A1/SULT1A1 Monoclonal specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 1A1/SULT1A1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 34
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 1A1/SULT1A1 (P50225-1; Glu2-Leu295).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6817
Test Specificity Has cross-reactivity in ELISA with: Human SULT1E1 No cross-reactivity in ELISA with: Human SULT2A1 Human SULT2B1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.