missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic beta-Glucosidase/GBA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99838-100ul
This item is not returnable.
View return policy
Description
Cytosolic beta-Glucosidase/GBA3 Polyclonal antibody specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for ELISA
Specifications
| Cytosolic beta-Glucosidase/GBA3 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878 | |
| Produced in rabbits immunized with purified, recombinant Human Cytosolic beta-Glucosidase/GBA3 (Accession#: NP_066024.1; Met 1-Leu 469) | |
| 100 μg | |
| metabolism | |
| 57733 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction