missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic beta-Glucosidase/GBA3 Antibody, FITC, Novus Biologicals™

Product Code. 18352786 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352786 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352786 Supplier Novus Biologicals Supplier No. NBP299838F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cytosolic beta-Glucosidase/GBA3 Polyclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic beta-Glucosidase/GBA3
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cytosolic beta-Glucosidase/GBA3 (NP_066024.1; Met 1-Leu 469).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57733
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.