missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Antibody (OTI8A2), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72019JF549
This item is not returnable.
View return policy
Description
Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Monoclonal specifically detects Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CNDP2 | |
| Full length recombinant protein of human Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL (NP_060705) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI8A2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1 | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 55748.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction