missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Antibody (210), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90495JF646
This item is not returnable.
View return policy
Description
Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Monoclonal specifically detects Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Has cross-reactivity in ELISA with: Human CNDP2 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 210 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL (NP_075638.2; Met1-Asn475). | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 55748 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction