missing translation for 'onlineSavingsMsg'
Learn More

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Antibody (1B1), Novus Biologicals™

Product Code. 18341619 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341619 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341619 Supplier Novus Biologicals Supplier No. H00055748M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Monoclonal antibody specifically detects Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 1B1
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060705
Gene Alias CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1
Host Species Mouse
Immunogen CNDP2 (NP_060705, 191 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VCISDNYWLGKKKPCITYGLRGICYFFIEVECSNKDLHSGVYGGSVHEAMTDLILLMGSLVDKRGNILIPGINEAVAAVTEEEHKLYDDIDFDIEEFAKDVGAQILLHSH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 55748
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.