missing translation for 'onlineSavingsMsg'
Learn More

Cytosine, TRC

Product Code. 30137910
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137910 5 g 5g
30133857 10 g 10g
17968394 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137910 Supplier Toronto Research Chemicals Supplier No. TRCC9989505G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-30-7
Molecular Formula C4 H5 N3 O
Molecular Weight (g/mol) 111.1
Synonym 2(1H)-Pyrimidinone, 6-amino-, 2(1H)-Pyrimidinone, 4-amino- (9CI), Cytosine (8CI), 6-Amino-2(1H)-pyrimidinone, 4-Amino-1H-pyrimidin-2-one, 4-Amino-2(1H)-pyrimidinone, 4-Amino-2-hydroxypyrimidine, 4-Amino-2-oxo-1,2-dihydropyrimidine, 4-Aminouracil, Cytosines, Cytosinimine, NSC 27787, 4-Aminopyrimidin-2(1H)-one, Cytosine, Gemcitabine Hydrochloride Imp. A (EP), Lamivudine Imp. E (EP)
IUPAC Name 4-amino-1H-pyrimidin-2-one
SMILES NC1=NC(=O)NC=C1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H5N3O/c5-3-1-2-6-4(8)7-3/h1-2H,(H3,5,6,7,8)
Formula Weight 111.0433
Percent Purity >95%
Chemical Name or Material Cytosine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.