missing translation for 'onlineSavingsMsg'
Learn More

Cytosine, MedChemExpress

Product Code. 16431248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431248 Supplier Medchem Express Supplier No. HYI0626
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cytosine is one of the four main bases found in DNA and RNA. Cytosine modifications exhibit circadian oscillations that are involved in epigenetic diversity and aging.

Cytosine is one of the four main bases found in DNA and RNA. Cytosine modifications exhibit circadian oscillations that are involved in epigenetic diversity and aging.

Specifications

Chemical Name or Material Cytosine
CAS 71-30-7
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C4H5N3O
Quantity 5 g
Solubility Information DMSO : 16.67 mg/mL (150.05 mM; Need ultrasonic)
SMILES O=C1NC=CC(N)=N1
Molecular Weight (g/mol) 111.1
Formula Weight 111.1
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.