missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosine-β-D-arabinofuranoside Hydrochloride, Fisher BioReagents
Click to view available options
Quantity:
100 mg
500 mg
5 g
Packaging:
Amber Glass
Description
Cytosine-β-D-Arabinofuranoside Hydrochloride inhibits incorporation of labeled thymidine into DNA.
Specifications
Specifications
Melting Point | 185°C to 195°C |
Color | White |
Packaging | Amber Glass |
Quantity | 5 g |
Formula Weight | 279.7 |
Physical Form | Powder |
Chemical Name or Material | Cytosine-β-D-Arabinofuranoside Hydrochloride |
Safety and Handling
Product Identifier
- Cytosine beta-D-arabinofuranoside hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H361d-Suspected of damaging the unborn child.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'