missing translation for 'onlineSavingsMsg'
Learn More

Cytosine-β-D-arabinofuranoside Hydrochloride, Fisher BioReagents

Product Code. 10012954
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
Packaging:
Amber Glass
Unit Size:
100mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10012954 500 mg Amber Glass 500mg
11483064 100 mg Amber Glass 100mg
10696195 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10012954 Supplier Fisher Bioreagents Supplier No. BP2512500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Cytosine-β-D-Arabinofuranoside Hydrochloride inhibits incorporation of labeled thymidine into DNA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-74-9
Molecular Formula C9H14ClN3O5
Molecular Weight (g/mol) 279.68
MDL Number MFCD00012839
InChI Key KCURWTAZOZXKSJ-JBMRGDGGSA-N
Synonym cytarabine hydrochloride
PubChem CID 134129637
IUPAC Name 4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 185°C to 195°C
Color White
Packaging Amber Glass
Quantity 500 mg
Formula Weight 279.7
Physical Form Powder
Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Cytosine beta-D-arabinofuranoside hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.