missing translation for 'onlineSavingsMsg'
Learn More

Cytosine beta-D-arabinofuranoside hydrochloride, 99%, crystalline, Thermo Scientific Chemicals

Catalog Number p-3778100
136.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
69-74-9
C9H14ClN3O5
279.68
MFCD00012839
KCURWTAZOZXKSJ-JBMRGDGGSA-N
cytarabine hydrochloride
134129637
Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
69-74-9
C9H14ClN3O5
279.68
MFCD00012839
KCURWTAZOZXKSJ-JBMRGDGGSA-N
cytarabine hydrochloride
134129637
Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 197.0°C to 198.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 10, 2778
Specific Rotation +127° to +133° (20°C, 589nm) ((c=0.7, H2O))
IUPAC Name 4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
Formula Weight 279.67
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Cytosine β-D-arabinofuranoside hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cytosine beta-D-arabinofuranoside hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'