missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytosine-beta-D-arabinofuranose hydrochloride

Product Code. p-7043132
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 15437029

Brand: Thermo Scientific Chemicals J65671.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interferes with the synthesis of DNA | C9H14ClN3O5 | 279.68 g/mol

Interferes with the synthesis of DNA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Molecular Weight (g/mol) 279.68
Product Type Cytosine-beta-D-arabinofuranose hydrochloride
Molecular Formula C9H14ClN3O5
MDL Number MFCD00012839
Quantity 1 g
Content And Storage Keep cold
InChI Key KCURWTAZOZXKSJ-JBMRGDGGSA-N
SMILES Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
Color White

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.