missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus pp65 Antibody (B051M) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68199
This item is not returnable.
View return policy
Description
Cytomegalovirus pp65 Monoclonal specifically detects Cytomegalovirus pp65 in Cytomegalovirus, Virus samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Cytomegalovirus pp65 | |
| Monoclonal | |
| 1.3 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| B051M | |
| Unconjugated | |
| 64 kDa lower matrix phosphoprotein, 65 kDa lower matrix phosphoprotein, CMV, CMV 65 kDa lower matrix phosphoprotein, CMV pp65, CMV pp65 IE, CMV tegument protein UL83, cytomegalovirus, Cytomegalovirus 65 kDa lower matrix phosphoprotein, Cytomegalovirus pp65, Cytomegalovirus tegument protein UL83, GP64, PP64, PP65, Tegument protein UL 83, Tegument protein UL83, UL 83, UL83 | |
| CMV Lysate | |
| 0.05 mg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Specific for pp65 of CMV. Functions in pp65 antigenemia assay (indirect IFA). (B051M) | |
| Store at 4degreesC. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction