missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus p65 Antibody (CMV100), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34595IR
This item is not returnable.
View return policy
Description
Cytomegalovirus p65 Monoclonal specifically detects Cytomegalovirus p65 in Cytomegalovirus, Virus samples. It is validated for ELISA.
Specifications
| Cytomegalovirus p65 | |
| Monoclonal | |
| DyLight 755 | |
| CMV, p65 | |
| Recombinant CMV-p65 protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| CMV100 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| 65 kDa | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction