missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cytomegalovirus gH Pentamer Complex Polyclonal antibody specifically detects Cytomegalovirus gH Pentamer Complex in Cytomegalovirus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Cytomegalovirus gH Pentamer Complex |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA |
| Formulation | PBS pH 7.4 |
| Gene Alias | envelope glycoprotein H |
| Host Species | Sheep |
| Immunogen | Recombinant CMV gH pentameric complex, expressed in mammalian cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?