missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytomegalovirus gH Late Antigen Recombinant Human Monoclonal Antibody (MSL-109)
GREENER_CHOICE

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA551801

Product Code. 30087407

  • £477.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Binds human cytomegalovirus (CMV).

Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytomegalovirus gH Late Antigen
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Human
Protein A
RUO
Virus
Antibody
IgG1 κ
Neutralization
MSL-109
Unconjugated
CMV
CMV antigen (Virus antigen batch V2547) prepared from MRC-5 cells infected with the Towne strain of CMV.
200 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.