missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytomegalovirus gH Late Antigen Chimeric Recombinant Rabbit Monoclonal Antibody (MSL-109)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551600
This item is not returnable.
View return policy
Description
Binds human cytomegalovirus (CMV).
Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells.
Specifications
| Cytomegalovirus gH Late Antigen Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG κ |
| ELISA, Neutralization | |
| MSL-109 | |
| Unconjugated | |
| CMV | |
| CMV antigen (Virus antigen batch V2547) prepared from MRC-5 cells infected with the Towne strain of CMV. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction