missing translation for 'onlineSavingsMsg'
Learn More

Cytomegalovirus gB Antibody, DyLight 594, Novus Biologicals™

Product Code. 18346785 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346785 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346785 Supplier Novus Biologicals Supplier No. NBP306076DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cytomegalovirus gB Polyclonal specifically detects Cytomegalovirus gB in Cytomegalovirus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus gB
Applications ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CMV, CMV glycoprotein B, CMV glycoprotein GP55, Cytomegalovirus glycoprotein GP55, gB, Glycoprotein B, Glycoprotein GP55, HCMV, HCMV glycoprotein B, HCMV glycoprotein GP55, HHV 5 glycoprotein B, HHV 5 glycoprotein GP55, HHV5 glycoprotein B, HHV5 glycoprotein GP55, Human herpesvirus 5 glycoprotein B, Human herpesvirus 5 glycoprotein GP55, UL55
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Cytomegalovirus gB (AAA45920.1) extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Cytomegalovirus (CMV)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.