missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus Antibody (HCMV16 (51C1)), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64398AF350
This item is not returnable.
View return policy
Description
Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Cytomegalovirus | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| HCMV16 (51C1) | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| Cytomegalovirus extracts | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| NB100-64398 is specific for Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells. It does not react with HSV, VZV, EBV or uninfected cells. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction