missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-65055-0.1ML
This item is not returnable.
View return policy
Description
Cytomegalovirus Polyclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Cytomegalovirus | |
| Polyclonal | |
| Unconjugated | |
| Phosphate buffered saline with 0.1% Sodium Azide | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| Goat | |
| IgG purified | |
| RUO | |
| Predominant reactivity is to glycoprotein B. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Frozen), ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 4.0-5.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P06473 | |
| gB | |
| Purified virons of strain AD169. | |
| 0.1 mL | |
| Primary | |
| Virus, Cytomegalovirus (CMV) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction