missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody (SPM115 + SPM116), Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £392.00
Specifications
| Antigen | Cytokeratin, pan |
|---|---|
| Clone | SPM115 + SPM116 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL, Flow (Intracellular) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18474852
|
Novus Biologicals
NBP2-34347-0.1mg |
0.1 mg |
£392.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18488731
|
Novus Biologicals
NBP2-34347-0.02mg |
0.02 mg |
£232.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)Specifications
| Cytokeratin, pan | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK | |
| Human epidermal keratin | |
| Primary | |
| Store at 4C. |
| SPM115 + SPM116 | |
| Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL, Flow (Intracellular) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit | |
| Q7Z794, Q01546 | |
| 3848 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title