missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody (PAN-CK (Cocktail)), PerCP, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cytokeratin, pan Monoclonal specifically detects Cytokeratin, pan in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | Cytokeratin, pan |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PAN-CK (Cocktail) |
| Conjugate | PerCP |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Symbols | KRT1 |
| Host Species | Mouse |
| Immunogen | This Cytokeratin, pan Antibody (PAN-CK (Cocktail)) was developed against human epidermal keratin. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?