missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody (K20) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £302.00
Specifications
| Antigen | Cytokeratin, pan |
|---|---|
| Clone | K20 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655296
|
Novus Biologicals
NBP2-50220-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695457
|
Novus Biologicals
NBP2-50220 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Cytokeratin, pan | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| PBS | |
| 3848 | |
| IgG2a κ | |
| Protein A purified |
| K20 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A,NEPPK | |
| This Cytokeratin, pan Antibody (K20) was developed against hair keratin. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title