missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin, pan Antibody (Cocktail PCK/4933R), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18331867 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331867 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331867 Supplier Novus Biologicals Supplier No. NBP314098AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin, pan
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Cocktail PCK/4933R
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Crude cytokeratin extract prepared from RT-4 and MCF-7 cells.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3848
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.