Learn More
Cytokeratin, Low Molecular Weight, Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.
Brand: Epredia MS-341-RQ
Description
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. The basic keratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. Members of acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with cell type, differentiation status and environment. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.Host Species: Mouse
Clone: AE1
Isotype: IgG1
Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Epitope: Not determined
Immunogen: Human epidermal Cytokeratin
Positive Control: Skin, Squamous cell CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
| Cytokeratin, Low Molecular Weight Ab-1 | |
| Monoclonal | |
| Prediluted | |
| Mouse | |
| 12 mL | |
| Cytoskeletal Protein | |
| Ready-to-Use |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| AE1 | |
| Unconjugated | |
| Human epidermal keratin | |
| IVD | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.