missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin, Low Molecular Weight, Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Brand:  Epredia MS-341-R7

 View more versions of this product

Product Code. 12641077

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. The basic keratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. Members of acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with cell type, differentiation status and environment. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: AE1

Isotype: IgG1

Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Human epidermal Cytokeratin

Positive Control: Skin, Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin, Low Molecular Weight Ab-1
Monoclonal
Prediluted
Mouse
7 mL
Cytoskeletal Protein
Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin)
AE1
Unconjugated
Human epidermal keratin
IVD
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.