missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, LMW Rabbit anti-Human, Clone: KRTL/4440R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07287-100ug
This item is not returnable.
View return policy
Description
Cytokeratin, LMW Monoclonal specifically detects Cytokeratin, LMW in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin, LMW | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39 | |
| Synthetic peptide corresponding to Cytokeratin, LMW residues within aa100-200 of Cytokeratin, LMW | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| KRTL/4440R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 374454 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction