missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, LMW Antibody (KRTL/4440R) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cytokeratin, LMW Monoclonal specifically detects Cytokeratin, LMW in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Cytokeratin, LMW |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | KRTL/4440R |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin : 1-2 ug/mL |
| Formulation | 10mM PBS |
| Gene Alias | Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?