missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, LMW Antibody (KRTL/1077), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44373-0.02MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Cytokeratin, LMW Monoclonal specifically detects Cytokeratin, LMW in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin, LMW | |
| Monoclonal | |
| 0.2mg/mL | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
| Q7Z794 | |
| KRT77 | |
| Recombinant human KRT77 protein | |
| 0.02 mg | |
| Primary | |
| This MAb recognizes the 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50, 48, 46, 45, and 40kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| KRTL/1077 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 374454 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction