missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, HMW Mouse, Clone: 34BE12, Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £393.00
Specifications
| Antigen | Cytokeratin, HMW |
|---|---|
| Clone | 34BE12 |
| Dilution | Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18104384
|
Novus Biologicals
NBP2-34340-0.02MG |
0.02 mg |
£232.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769923
|
Novus Biologicals
NBP2-34340-0.1mg |
0.1 mg |
£393.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin, HMW Monoclonal specifically detects Cytokeratin, HMW in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin, HMW | |
| Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
| KRT76 | |
| IgG1 κ | |
| Protein A purified | |
| This antibody recognizes CK1, CK5, CK10 and CK14. In normal epithelia, it stains stratified epithelia, myoepithelial cells and basal cells in the prostate gland and bronchi. This MAb shows no reactivity with hepatocytes, pancreatic acinar cells, proximal renal tubules, or endometrial glands; there is no reactivity with cells derived from simple epithelia. Mesenchymal tumors, lymphomas, melanomas, neural tumors, and neuroendocrine tumors are negative with this antibody. It stains myoepithelial cells and has been shown to be useful in distinguishing prostate adenocarcinoma from benign prostate. This antibody has also been useful in separating benign from malignant intraductal breast proliferations. |
| 34BE12 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P04264 | |
| 51350 | |
| Solubilized keratin extract from human stratum corneum | |
| Primary | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title