missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin, HMW Antibody (AE-3), Novus Biologicals™

Product Code. 18424112 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
20 ug
Unit Size:
0.10mg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424112 20 ug 20µg
18196032 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18424112 Supplier Novus Biologicals Supplier No. NBP22940420ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

Cytokeratin, HMW Monoclonal specifically detects Cytokeratin, HMW in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin, HMW
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone AE-3
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/mL, Flow Cytometry 0.5-1 ug/million cells, ELISA 1-5 ug/mL for coating, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 0.5-1 ug/mL, Immunohistochemistry-Frozen 0.5-1 ug/mL
Gene Accession No. Q01546
Gene Alias CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9
Gene Symbols KRT76
Host Species Mouse
Immunogen Human epidermal keratin (Uniprot: Q01546)
Purification Method Protein A or G purified
Quantity 20 ug
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51350
Test Specificity This monoclonal antibody recognizes basic (Type II or HMW) cytokeratins, which include 67kDa (CK1); 64kDa (CK3); 59kDa (CK4); 58kDa (CK5); 56kDa (CK6); 52kDa (CK8). Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40kDa. monoclonal antibody AE3 recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE1/AE3 is a broad spectrum anti pan-keratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer.
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Rabbit
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.