Learn More
Invitrogen™ Cytokeratin 8 Monoclonal Antibody (M20)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA106318
Description
MA1-06318 detects cytokeratin 8 in rat, human, and rabbit samples. MA1-06318 has sucessfully been used in Western blotting, immunocytochemistry, flow cytometry and immunohistochemistry (frozen, paraffin). The MA1-06318 immunogen is keratin isolated from the human breast carcinoma cell line MCF-7. Store at 4°C or in small aliquots at -20°C.
Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
Specifications
| Cytokeratin 8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P05787, Q10758 | |
| KRT8 | |
| Keratin isolated from the human breast carcinoma cell line MCF-7. | |
| 100 μL | |
| Primary | |
| Human, Rat, Rabbit | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| M20 | |
| Unconjugated | |
| KRT8 | |
| AA960620; AL022697; AU019895; CARD2; cb186; CK8; CK-8; CYK8; CYKER; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; Cytokeratin-8; cytokeratin-A; DreK8; EndoA; etID33550.23; etID43142.23; etID43142.8; K2C8; K8; KERA; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 8; keratin, type 2, gene 8; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; keratin-8; KO; Krt-2.8; Krt2-8; krt8; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8; sb:cb186; type II cytokeratin A (no.8); type-II keratin Kb8; wu:fa20h05; wu:fa95h10; wu:fb96c06; zfk8; zf-K8 | |
| Mouse | |
| Purified | |
| RUO | |
| 100348891, 25626, 3856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.