missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8 Antibody (AE3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
£171.00 - £289.00
Specifications
| Antigen | Cytokeratin 8 |
|---|---|
| Clone | AE3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen |
| Applications | Immunohistochemistry (Frozen), Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18226742
|
Novus Biologicals
NB120-9287-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717083
|
Novus Biologicals
NB120-9287 |
£289.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Cytokeratin 8 Monoclonal specifically detects Cytokeratin 8 in Human, Mouse, Rat, Bovine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Cytokeratin 8 | |
| 1.0 mg/mL | |
| Immunohistochemistry (Frozen), Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| KRT8 | |
| IgG1 | |
| Affinity Purified | |
| Recognizes most of the basic (Type II) keratins, HMW cytokeratins. It is broadly reactive and stains positively almost all epithelia and their neoplasms. AE3 reacts with all layers of epidermis (basal layer and above). The antibody is an excellent marker for distinguishing carcinomas from non-epithelial tumours. It has been used for studying the immunolocalisation of epidermal keratins, tissue distribution of keratins, various diseases of epidermis, oral mucosa and other epithelia and other epithelia development. |
| AE3 | |
| Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Mouse | |
| Mouse, Primate, Rabbit, Chicken, Bovine, Rat, Human | |
| 3856 | |
| Human epidermal cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title