missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8/18 Rabbit anti-Human, Clone: KRT8.18/2297R, Novus Biologicals™
Rabbit Monoclonal Antibody
£235.00
Specifications
| Antigen | Cytokeratin 8/18 |
|---|---|
| Clone | KRT8.18/2297R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 8/18 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 3856 | |
| IgG | |
| Protein A or G purified |
| KRT8.18/2297R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| Recombinant full-length human Cytokeratin 8/18 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title