missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8/18 Mouse anti-Human, Clone: rKRT8.18/1346, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07851-100ug
This item is not returnable.
View return policy
Description
Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 8/18 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| Recombinant full-length human KRT8 protein and Recombinant full-length human KRT18 protein | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
| 3856 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rKRT8.18/1346 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction