missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8/18 Antibody (C-51) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48179
This item is not returnable.
View return policy
Description
Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human, Rat, Porcine, Bovine, Ferret (Negative), Monkey, Sheep, Chicken (Negative), Donkey (Negative), Mouse (Negative), Rabbit (Negative), Xenopus (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 8/18 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| KRT8 | |
| Cytoskeleton preparation from HeLa cells | |
| 7 mL | |
| Cancer, Cytoskeleton Markers | |
| 3856 | |
| Human, Rat, Pig, Ferret (Negative), Primate, Monkey, Sheep, Chicken (Negative), Donkey (Negative), Mouse (Negative), Rabbit (Negative), Xenopus (Negative) | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| C-51 | |
| Immunohistochemistry-Paraffin | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This monoclonal antibody recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 18 in the normal epidermis so only Paget cells are stained. | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction