missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 71 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86614-0.1ML
This item is not returnable.
View return policy
Description
Cytokeratin 71 Polyclonal specifically detects Cytokeratin 71 in Human samples. It is validated for Western Blot.
Specifications
| Cytokeratin 71 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 112802 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| CK-71, cytokeratin-71, hK6irs, hK6irs1, K6IRS1MGC119390, K71, KB34, Keratin 6 irs, keratin 71, keratin, type II cytoskeletal 71, Keratin-71, KRT6IRS, KRT6IRS1MGC119391, Type II inner root sheath-specific keratin-K6irs1, Type-II keratin Kb34 | |
| The immunogen is a synthetic peptide directed towards the middle region of human Cytokeratin 71 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction