missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 7 Recombinant Rabbit Monoclonal Antibody (23GB2915), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Cytokeratin 7 |
|---|---|
| Clone | 23GB2915 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089921
|
Invitrogen™
MA553274 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088027
|
Invitrogen™
MA553275 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 7 Recombinant Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Cytokeratin 7 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P08729 | |
| 3855 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2915 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729} | |
| A synthesized peptide derived from human Cytokeratin 7 (12-48AA). | |
| Primary | |
| -20°C | |
| KRT7 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title