missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cytokeratin 7 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
Specifications
Specifications
| Gene ID (Entrez) | 3855 |
| Protein Family | Cytokeratin 7 |
| Purity | >80% SDS-PAGE |
| Content And Storage | Store at -20 degrees C. Avoid freeze/thaw cycles. |
| Formulation | PBS, 1 M Urea, pH 7.4. |
| Gene Symbol | KRT7 |
| Quantity | 100 μg |
| Research Category | Cell Biology, Cellular Markers |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-55071. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto