missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7 Rabbit anti-Human, Clone: OV-TL 12/30, Novus Biologicals™
Rabbit Monoclonal Antibody
£210.00 - £463.00
Specifications
| Antigen | Cytokeratin 7 |
|---|---|
| Clone | OV-TL 12/30 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364062
|
Novus Biologicals
NBP3-12014-0.025MG |
0.025 mg |
£210.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18386113
|
Novus Biologicals
NBP3-12014-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| Cytokeratin 7 | |
| 1 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS | |
| 3855 | |
| IgG κ | |
| Protein A purified |
| OV-TL 12/30 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| This antibody was originally raised by immunizing BALB/c mice with ovarian carcinoma cell line OTN11. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title