missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582291
This item is not returnable.
View return policy
Description
Immunogen sequence: RQEESEQIKT LNNKFASFID KVRFLEQQNK LLETKWTLLQ EQKSAKSSRL PDIFEAQIAG LRGQLEALQV DGGRLEAELR SMQDVVEDFK NKYEDEINRR TAAENEFVVL KKDVD Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
Specifications
| Cytokeratin 7 | |
| Polyclonal | |
| Unconjugated | |
| KRT7 | |
| CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110310, 300242, 3855 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08729, Q6IG12, Q9DCV7 | |
| KRT7 | |
| Recombinant protein corresponding to Human KRT7. Recombinant protein control fragment (Product #RP-88684). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction